Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Hepa-E1

99072812, eel liver, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Hepa-E1, 99072812

biological source

eel liver

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Eel liver

Cell Line Description

The hepatocyte cell line Hepa-E1 was derived from water eel. The cells show no contact inhibition and have been reported to show apoptosis in the presence of actinomycin D, humic acid or TGF-β 1. Normal culture conditions are at 26-28°C, but maintenance at 4°C for up to 400 days was described as well.

Culture Medium

E-RDF (Kyokuto Pharmaceuticals)+ 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:8 using 0.25% trypsin or trypsin/EDTA 26-28°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service