Skip to Content
MilliporeSigma
All Photos(1)

Documents

L929 Cell Line from mouse

85011425, mouse adipose tissue, Fibroblast

Synonym(s):

Earles′s Cells, L Cells, NCTC clone 929 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

L929 Cell Line from mouse, mouse C3H/An connective tisue, 85011425

biological source

mouse adipose tissue

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C3H/An connective tissue

Cell Line Description

Subclone of parental strain L, established by W R Earle in 1940. One of the first to be established in continuous culture. The L strain was derived from normal subcutaneous areolar and adipose tissue of a 100 day old male C3H/An mouse. Cells are APRT+ and HPRT+.

Application

Virus studies: PRV, VSV (Indiana), HSV

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures seeding at 5x1,000 to 2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service