Skip to Content
MilliporeSigma
All Photos(1)

Documents

NCI-H69/LX10

96042331, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCI-H69/LX10, 96042331

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human small cell lung cancer, drug-resistant

Cell Line Description

The drug-resistant cell line NCI-H69/LX10 has been derived from the parent line NCI-H69 (ECACC No 91091802). It is recommended to culture the cells without drug upon resuscitation; add doxorubicine once growth is fully established.

Application

Drug-resistance studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 1.0 μg/ml doxorubicine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2, 37ºC; resuscitate in drug-free medium and add drug after growth has been established.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service