Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NCI-H69VCR/R

96042330, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

product name

NCI-H69VCR/R, 96042330

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human small cell lung cancer, drug-resistant

Cell Line Description

NCI-H69 VCR/R has been derived from the parent small cell lung cancer line NCI-H69 (Sigma Catalogue number. 91091802) It is recommended to culture cells without drug upon resuscitation; add vincristine once growth is fully established. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

Application

Drug-resistance studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 12
D16S539: 11
D5S818: 11
D7S820: 9
THO1: 8,9
TPOX: 10
vWA: 16,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 2.0 μg/ml vincristine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x 100,000 cells/ml; 5% CO2; 37ºC. Resuscitate in drug-free medium, add after growth has been established.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service