Skip to Content
MilliporeSigma
All Photos(1)

Documents

PC-9 (formerly known as PC-14)

90071810, human lung, Heterogeneous mixture of round cells and spindle shaped cells

Synonym(s):

PC-9/S1 Cells, PC-9S1 Cells, PC9 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

PC-9 (formerly known as PC-14), 90071810

biological source

human lung

growth mode

Adherent/suspension

karyotype

Not specified

morphology

Heterogeneous mixture of round cells and spindle shaped cells

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human adenocarcinoma

Cell Line Description

Derived from a human adenocarcinoma from lung tissue which remains undifferentiated.

PC-14 was originally deposited with the Riken BioResource Center as a cell line derived from human lung adenocarcinoma (undifferentiated type) in 1989. The RIKEN BioResource Center subsequently informed the European Collection of Cell Cultures (ECACC) that short tandem repeat (STR) DNA profile analysis, undertaken in collaboration Immuno-Biological Laboratories Co., Ltd Japan, has shown this cell line to be identical to PC-9 a cell line derived from human lung adenocarcinoma (differentiated type). The misidentification occurred prior to the cell line being deposited at the RIKEN BioResource Center. The name of the cell line has been changed to PC-9 to reflect this finding.

Application

PC-9 cell line has been used to study the effects of α-tocopherol addition on various cytotoxic drugs.
Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 8
D16S539: 9
D5S818: 11
D7S820: 10
THO1: 7
TPOX: 11
vWA: 17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Seed cells at 2-4 x 10,000 cells/cm2; 5% CO2; 37°C. Remove attached cells by brief treatment with 0.25% trypsin or trypsin/EDTA.  

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Related Content

Authenticated lung cancer cell lines cater to drug response, toxicology, and in vitro model needs, supporting various research applications.

Authenticated lung cancer cell lines cater to drug response, toxicology, and in vitro model needs, supporting various research applications.

Authenticated lung cancer cell lines cater to drug response, toxicology, and in vitro model needs, supporting various research applications.

Authenticated lung cancer cell lines cater to drug response, toxicology, and in vitro model needs, supporting various research applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service