Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OV17R

96020763, human ovary, Epithelial

Synonym(s):

Human OV17R, OV17R Cell Line, OV17R Human Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

OV17R, 96020763

biological source

human ovary

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human ovarian adenocarcinoma (ascites) stage III

Cell Line Description

The OV17R cell line was derived from a human ovarian adenocarcinoma (ascites) stage III of a 50 year-old female.

Culture Medium

DMEM:HAMS F12 (1:1) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS) + 0.4 ug/ml hydrocortisone + 10ug/ml insulin

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service