Skip to Content
MilliporeSigma
All Photos(1)

Documents

COV644

7071908, human ovary, Polygonal & fusiform

Synonym(s):

COV-644 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COV644, 07071908

biological source

human ovary

growth mode

Adherent

karyotype

Modal chromosome no. 58 (50-60)

morphology

Polygonal & fusiform

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human ovarian epithelial-mucinous carcinoma, primary tumour

Cell Line Description

Human ovarian epithelial-mucinous carcinoma cell line established from a solid primary tumour. Anchorage-independent growth in agar has been reported.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split confluent cultures 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service