Skip to Content
MilliporeSigma
All Photos(1)

Documents

OAW42

85073102, human ovary, Epithelial

Synonym(s):

OAW-42 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

OAW42, 85073102

biological source

human ovary

growth mode

Adherent

karyotype

2n = 86

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human ovarian tumour epithelial

Cell Line Description

From an ascites of patient with ovarian cystadenocarcinoma. Has retained ability to form free floating cysts in vitro and produces extracellular matrix. Has defined chemosensitivity pattern. Valuable for studies on biology of human ovarian cancer.

Application

OAW42 has been used to investigate the cytotoxic potential of histone deacetylase 1 as a target for ovarian cancer immunotherapy and to study the effects of tazemetostat, a potent and selective EZH2 inhibitor, on OAW42 cell lines.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11
D16S539: 12,13
D5S818: 11,12
D7S820: 8
THO1: 6,7
TPOX: 8,11
vWA: 15,16

Culture Medium

DMEM + 2mM Glutamine + 1mM Sodium Pyruvate (NaP) + 20 IU/l Bovine Insulin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service