Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COV413A

7071905, human ovary (metastasis), Polygonal & fusiform

Synonym(s):

COV 413A Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COV413A, 07071905

biological source

human ovary (metastasis)

growth mode

Adherent

karyotype

Not specified

morphology

Polygonal & fusiform

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human ovarian epithelial-serous carcinoma, sigmoid

Cell Line Description

A human ovarian epithelial-serous carcinoma cell line established from a metastasis of the sigmoid colon. This cell line was derived from the same patient as the ovarian cell line COV413B (ECACC cat no. 07071906).

Culture Medium

DMEM + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Split confluent cultures 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service