Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TK6

95111735, human lymph, Not specified

Synonym(s):

H2BT Cells, TK-6 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

TK6, 95111735

biological source

human lymph

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lymphoblast, thymidine kinase heterozygote

Cell Line Description

The thymidine kinase heterozygote cell line TK6 has been isolated from the lymphoblastoid line HH4. HH4 was derived from the WIL-2 cell line. TK6 cells are CD19 positive, 50% of the cell population express CD20 and a small sub-population has been tested positive for CD22. We cannot confirm the BVDV status of this cell line.

Application

Bromodeoxyuridine resistant assays
TK-6 cell line has been used to study the in vitro genotoxic effects of trans- and cis-2-ethylhexyl 4-methoxycinnamate (EHMC). It has also been used to study the interaction of silver nanomaterials with the cells.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 11
D16S539: 11,12
D5S818: 12,13
D7S820: 9,11
THO1: 8,9.3
TPOX: 8,11
vWA: 17,20

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Horse Serum (HS) or FBS.

Subculture Routine

Maintain cultures between 1-9 x 100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service