Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

J774A.1 Cell Line from mouse

91051511, mouse blood, Not specified

Synonym(s):

J774 A1 Cells, J774.A1 Cells, J774A1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

J774A.1 Cell Line from mouse, 91051511

biological source

mouse blood

description

Mouse BALB/c monocyte macrophage

growth mode

Semi-adherent

karyotype

Not specified

morphology

Not specified

products

Interleukin 1 (IL-1), lysozyme

receptors

Immunoglobulin, complement

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c monocyte macrophage

Cell Line Description

Derived from a tumour in a female BALB/c mouse. Cell line produces IL-5 and lysozyme.

Application

J774A.1 cell line has been used to test the cytotoxicity of [1,2,3]triazolo[1,5-a]pyridinium salts (having leishmanicidal activity) from triazolopyridines. It has also been used to determine the antitrypanosomatid activity of flavonoid glycosides isolated from Delphinium sps.
Metabolic studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Cells are subcultured by harvesting attached and suspension cells. Remove majority of medium and centrifuge to harvest cells growing in suspension. Scrape adherent cells remaining in the flask. Resuspend cell pellet from centrifugation and combine with scraped cells. Seed into new flask at 1:3 to 1:4 split ratio i.e. seeding at 1-3 x 10,000 cells/cm2 Incubate at 37°C; 5% CO2. Please Note :- On receipt growing order cells may be in suspension. The cells and media should be removed from the flask and spun down to form a pellet. This should then be re-suspended in fresh media and a cell count performed. The cells should then be reseeded at 2-4 x 10E4/cm and should begin to show signs of re-attachment within 48 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service