Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MDH-2N1

Millipore

Millicell® Disposable Hemocytometer

Neubauer Improved ruling pattern, 2 chambers/slide, 10 µL loading volume, individually wrapped

Synonym(s):

Optical grade plastic hemocytometer, disposable hemocytometer, Cell Culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161502
NACRES:
NC.56

material

colorless plastic

sterility

non-sterile

feature

pyrogen free

packaging

pack of 50 (individually wrapped)

parameter

10 μL sample volume

technique(s)

cell counting: suitable

W × L × H

25 mm × 75 mm × 1.6 mm

chamber depth

0.1 mm

size

2 wells

Looking for similar products? Visit Product Comparison Guide

General description

The Millicell® disposable hemocytometer is used to manually count cells. It consists of a surface-patterned two enclosed chambers with two ports for sample injection. Count cells with the in the same way as you do with the Neubauer Improved counting chamber.

The Neubauer Improved counting chamber consists of 9 large squares, each measuring 1 x 1 mm. Each square has a total volume of 0. 1 mm3. The cells are distributed over 3 x 3 large squares, each with 1 mm edge length and with a surface area of 1 mm2.

With the Millicell® disposable hemocytometer, you inject 10 µL of sample, stained or unstained, into the desired chamber.

Application

cell counting, cell culture, hemocytometer

Features and Benefits

  • Quartz grade optical plastic
  • No cover slips
  • No washing

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Data for validated non-human cell lines using the Millicell® DCI Digital Cell Imager for confluency determination and estimated cell counts.

Data for validated non-human cell lines using the Millicell® DCI Digital Cell Imager for confluency determination and estimated cell counts.

Cell validation data using the Millicell® DCI Digital Cell Imager for confluency determination and estimated cell counts.

Cell validation data using the Millicell® DCI Digital Cell Imager for confluency determination and estimated cell counts.

See All

Related Content

Monitor your cell cultures with ease with the Millicell® DCI Digital Cell Imager. From confluency to cell count and morphology, the Millicell® DCI automates cell culture measurements for quicker, more consistent culturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service