Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HCT-8

90032006, human intestine, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HCT-8, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 90032006

biological source

human intestine

packaging

tube of 5 μg 90032006-DNA-5UG
pkg of vial of cells 90032006-1VL

growth mode

Adherent

karyotype

2n = 46, pseudodiploid

morphology

Epithelial

products

Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human ileocecal adenocarcinoma

Cell Line Description

From a 67 year old male. The four cell lines: DLD-1 (90102540), HCT-15 (91030712), HCT-8 (90032006) and HRT-18 (86040306) have been shown to have a common genetic origin.

Application

Tumourigenicity studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 1mM Sodium Pyruvate (NaP) + 10% Horse Serum (HS) or 5% Horse Serum (HS) + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service