Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HCT 116 Cell Line human

91091005, human colon, Epithelial-like

Synonym(s):

HCT116 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HCT 116 Cell Line human, 91091005, colon carcinoma

biological source

human colon

growth mode

Adherent

karyotype

Modal no. 45

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon carcinoma

Cell Line Description

One of 3 strains of malignant cells isolated from a male with colonic carcinoma. Cells are tumourigenic in nude mice and form colonies on agarose. Growth and plating efficiency are enhanced by using a feeder layer of murine fibroblasts.

Application

HCT 116 Cell Line has been used to:
  • study the importance of cyclin D1 for the activity of lithocholic acid hydroxyamide (LCAHA)
  • study the effects of different amounts of transferrin within the polyethyleneimine (PEI)-small interfering RNA (siRNA) complexes on the uptake of fluorescently-labelled siRNA
  • determine the effect of PTS (polyamine transport system) in the selective uptake of Quilamine HQ1-44, an iron chelator
Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 7,10
D13S317: 10,12
D16S539: 11,13
D5S818: 10,11
D7S820: 11,12
THO1: 8,9
TPOX: 8
vWA: 17,22

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service