Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HT29-MTX-E12

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 12040401, human colon, Epithelial

Synonym(s):

HT-29-MTX-E12 Cells, HT29, HT29 Cells, HT29-MTX, HT29-MTX Cells, MTX-E12 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human colon

packaging

tube of 5 μg 12040401-DNA-5UG
pkg of vial of cells 12040401-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

HT29, HT29-MTX, methotrexate, mucous layer, intestinal permeation, oral drug absorption

Cell Line Description

HT29 cells were differentiated into mature goblet cells using methotrexate. Mucous-secreting HT29-MTX subclones were isolated from this cell clone and characterized with regard to tight junction formation, development of confluent monolayers and productio

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 11,12
D13S317: 11
D16S539: 11,12
D5S818: 11,12
D7S820: 10
THO1: 6
TPOX: 8,9
vWA: 17

Culture Medium

DMEM + 2mM Glutamine + 1% Non essential amino acids + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split subconfluent cultures (70-80%) 1:3 to 1:6 using 0.25% trypsin/EDTA; 7.5-10% CO2; 37 °C. Suggested seeding density 2-4x10,000 cells/cm².

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service