Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TM4

88111401, mouse testis, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

TM4, 88111401

biological source

mouse testis

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Transferrin, H-Y Ag, novel retinol-binding protein

receptors

Androgen, oestrogen and progesterone receptors

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c testis sertoli cell

Cell Line Description

From primary culture of sertoli cell enriched preparations from normal testes of a 11-13 day old BALB/c nu/+ mice. Cells respond to FSH but not LH; Plasminogen activator secretion is stimulated by FSH and also retinoic acid. Note: only distributed frozen.

Application

Hormone studies

Culture Medium

Ham′s F12 + DMEM (1:1) + 2mM Glutamine + 5% Horse Serum (HS) + 2.5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:20 to 1:200 i.e. seeding at 1x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Cells resuscitated from frozen stock should be split 1:3 at the first passage and thereafter as above.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service