Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GS-109-V-20

90110505, human skin, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

GS-109-V-20, 90110505

biological source

human skin

growth mode

Adherent

karyotype

2n = 46

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian skin fibroblast Gardner′s syndrome

Cell Line Description

Derived from skin from the upper arm of an 18 year old Caucasian male with Gardner′s syndrome is an autosomal dominant condition which predisposes to carcinoma and multiple polyps of the colon. Cells exist in an initiated state rather than a normal or transformed state. Normal diploid karyotype.

Application

Tumourigenicity studies

Culture Medium

EMEM (HBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures 1:2 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. Monolayer will form in a 25cm2 flask in 1 week.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service