Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TM3

91060526, mouse testis, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

TM3, 91060526

biological source

mouse testis

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Prostaglandin F2a

receptors

Epidermal growth factor (EGF), Androgen, Oestrogen, Progesterone

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c testis, Leydig cell

Cell Line Description

Originated from primary cultures of Leydig cell enriched preparations from normal testes of 11-13 day old BALB/c nu/+ mice. Characteristics similar to primary cultures of Leydig cells. Responds to leuteinising hormone but not FSH. In the presence of LH the cells are capable of metabolising cholesterol. These cells are non-tumourigenic.

Application

Hormone studies

Culture Medium

Ham′s F12 + DMEM (1:1) + 2mM Glutamine + 5% Horse Serum (HS) + 2.5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:200 i.e. seeding at 1x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37²C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service