Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCC207

Sigma-Aldrich

INS-1 832/13 Rat Insulinoma Cell Line

Rat

Synonym(s):

INS1 832/13, INS-1 (832/13), INS 832/13, 832/13, 832/13 INS-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

product name

INS-1 832/13 Rat Insulinoma Cell Line, INS-1 832/13 rat insulinoma cell line is a useful model for insulin secretion regulation and pancreatic islet beta-cell function studies.

biological source

rat

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

General description

Glucose-stimulated insulin secretion (GSIS) is potentiated by pancreatic beta cells and is critical to the physiological control of blood glucose levels. Insulin secretion is impaired in type 2 diabetes, and insight into the mechanisms and regulation of insulin secretion is fundamental to understanding the roles of beta cells in metabolic disease.

Cell Line Description

Cancer Cells

Application

INS-1 832/13 rat insulinoma cell line is a useful model for insulin secretion regulation and pancreatic islet beta-cell function studies.
Research Category
Metabolism
Research Sub Category
Diabetes
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Quality

• Each vial contains ≥ 1X10⁶ viable cells.
• Cells are tested negative for infectious diseases by a Mouse/Rat Comprehensive CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are verified to be of rat origin and negative for inter-species contamination from mouse, chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immacolata Prevenzano et al.
EMBO reports, 23(7), e52990-e52990 (2022-05-28)
Tight control of glycemia is a major treatment goal for type 2 diabetes mellitus (T2DM). Clinical studies indicated that factors other than poor glycemic control may be important in fostering T2DM progression. Increased levels of methylglyoxal (MGO) associate with complications
Daniela G Dengler et al.
Biochemical pharmacology, 185, 114451-114451 (2021-02-06)
The secretin receptor (SCTR) is a prototypic Class B1 G protein-coupled receptor (GPCR) that represents a key target for the development of therapeutics for the treatment of cardiovascular, gastrointestinal, and metabolic disorders. However, no non-peptidic molecules targeting this receptor have
Feng Li et al.
Cell reports, 42(1), 111904-111904 (2023-01-21)
TEAD1 and the mammalian Hippo pathway regulate cellular proliferation and function, though their regulatory function in β cells remains poorly characterized. In this study, we demonstrate that while β cell-specific TEAD1 deletion results in a cell-autonomous increase of β cell
Fabian Wirth et al.
Nature communications, 14(1), 6294-6294 (2023-10-10)
In patients with type 2 diabetes, pancreatic beta cells progressively degenerate and gradually lose their ability to produce insulin and regulate blood glucose. Beta cell dysfunction and loss is associated with an accumulation of aggregated forms of islet amyloid polypeptide
H Bischof et al.
Current protocols in chemical biology, 11(3), e71-e71 (2019-09-05)
Vital cells maintain a steep potassium ion (K+ ) gradient across the plasma membrane. Intracellular potassium ion concentrations ([K+ ]) and especially the [K+ ] within the extracellular matrix are strictly regulated, the latter within a narrow range of ∼3.5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service