Skip to Content
MilliporeSigma
All Photos(1)

Documents

LLC-PK1

86121112, Porcine kidney, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

LLC-PK1, 86121112

biological source

Porcine kidney

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Plasminogen activator

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Porcine kidney

Cell Line Description

Cells have been used in the production of plasminogen activator.

Application

Production of plasminogen activator

Culture Medium

Medium 199 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service