Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0766

Sigma-Aldrich

GSK343

≥98% (HPLC)

Synonym(s):

N-[(1,2-Dihydro-6-methyl-2-oxo-4-propyl-3-pyridinyl)methyl]-1-(1-methylethyl)-6-[2-(4-methyl-1-piperazinyl)-4-pyridinyl]-1H-indazole-4-carboxamide, N-[(6-Methyl-2-oxo-4-propyl-1,2-dihydropyridin-3-yl)methyl]-6-[2-(4-methylpiperazin-1-yl)pyridin-4-yl]-1-(propan-2-yl)-1H-indazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H39N7O2
CAS Number:
Molecular Weight:
541.69
MDL number:
UNSPSC Code:
41106514
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, white to beige to brown

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(CC1)CCN1C2=NC=CC(C3=CC4=C(C=NN4C(C)C)C(C(NCC5=C(CCC)C=C(C)NC5=O)=O)=C3)=C2

InChI

1S/C31H39N7O2/c1-6-7-23-14-21(4)35-31(40)26(23)18-33-30(39)25-15-24(16-28-27(25)19-34-38(28)20(2)3)22-8-9-32-29(17-22)37-12-10-36(5)11-13-37/h8-9,14-17,19-20H,6-7,10-13,18H2,1-5H3,(H,33,39)(H,35,40)

InChI key

ULNXAWLQFZMIHX-UHFFFAOYSA-N

Application

GSK343 has been used:
  • as an inhibitor of enhancer of zeste homolog 2 (EZH2) to assess the impact of chromatin condensation on cell migration
  • as histone methyl transferase inhibitor, to suppress the Polyandrocarpa misakiensis, mitochondrial non-coding-region (NCR) PmNCR reporter gene expression, facilitated by TC14-3
  • as a supplement in the growth medium to inhibit the generation of H3K27me3 or to inhibit RNA polymerase II
  • as EZH2 inhibitor to treat castration-resistant neuroendocrine prostate cancer (CRPC-NE)

Biochem/physiol Actions

GSK343 is a potent, specific inhibitor of the histone H3-lysine 27 (H3K27) methyltransferase EZH2. GSK343 inhibits EZH2 enzymatic activity with an IC50 of 4 nM. The compound displays 60 fold selectivity for EZH2 vs. EZH1, and 1000 fold or greater selectivity against other histone methyltransferases. The IC50 for inhibition of H3K27 methylation is < 200 nM in HCC1806 cells. For full characterization details, please visit the GSK343 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

GSK343 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

GSK343 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the GSK343 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patient derived organoids to model rare prostate cancer phenotypes
Puca L, et al.
Nature Communications, 9(1), 2404-2404 (2018)
Regulatory cis-and trans-elements of mitochondrial D-loop-driven reporter genes in budding tunicates
Kawamura K, et al.
Mitochondrion, 35, 59-69 (2017)
N A Wijetunga et al.
Oncogene, 36(14), 2030-2044 (2016-10-11)
The predisposition of patients with Hepatitis C virus (HCV) infection to hepatocellular carcinoma (HCC) involves components of viral infection, inflammation and time. The development of multifocal, genetically distinct tumours is suggestive of a field defect affecting the entire liver. The
Increased chromatin plasticity supports enhanced metastatic potential of mouse melanoma cells
Maizels Y, et al.
Experimental Cell Research, 357(2), 282-290 (2017)
Zhi-Chang Zhang et al.
Journal of Cancer, 9(1), 71-80 (2018-01-02)
Osteosarcoma (OS), which affects adolescents especially during a growth spurt, has the highest incidence of any primary malignant bone tumour, and a high rate of early metastasis leading to a very poor prognosis. In recent years, non-coding RNAs, especially long

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service