Skip to Content
MilliporeSigma
All Photos(1)

Documents

MEF-1

98061101, mouse embryo, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MEF-1, 98061101

biological source

mouse embryo

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse embryo fibroblast, SV40 - transformed

Cell Line Description

The cell line MEF-1 was established by transfection of mouse embryo fibroblasts ((C57 BL/6 x 129) F1) with a SV40 coding plasmid. MEF-1 express the wild-type for low density lipoprotein receptor related protein (LRP). Together with the cell lines PEA10 (Sigma Catalogue number. 97040310) and PEA13 (Sigma Catalogue number. 97040311), which express heterozygous and homozygous forms of LPR, an experimental system is available for the analysis of cellular uptake of functionally diverse ligands and the effect of LPR - deficiency. This is of importance since LRP - deficient mouse embryos die early during gestation.

Application

Study of LRP and LRP-deficiency

Culture Medium

DMEM + 2 mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:6 to 1:10 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service