Skip to Content
MilliporeSigma
All Photos(1)

Documents

L929/R

4102001, mouse adipose tissue, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

L929/R, 04102001

biological source

mouse adipose tissue

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C3H/An connective tissue cisplatin resistant

Cell Line Description

This cisplatin-resistant cell line has been developed by exposure of the parent L929 murine fibroblast cell line (Sigma Catalogue number. 85011425) to increasing concentrations of cisplatin in vitro. L929/R cells can be used in the development of novel anti-cancer treatments. The parent cell line L929 was derived from normal subcutaneous areolar adipose tissue.

Application

Development of novel anti-cancer treatments

Culture Medium

RPMI 1640 + 2mM L-Glutamine + 10% Foetal Bovine Serum (FBS) + 0.26μM Cisplatin.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:50 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 2% CO2; 37 C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service