Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NS0-Serum-Free Cell Line from mouse

3061601, mouse blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NS0-Serum-Free Cell Line from mouse, 03061601

biological source

mouse blood

description

Mouse myeloma, serum-free

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse myeloma, serum-free

Cell Line Description

NS0 cell line (Sigma Catalogue no. 85110503) adapted to grow in the serum free medium EX-CELL® NS0 Serum-Free Medium for NS0 Cells (Sigma 14650C). Suitable for use as a fusion parent for serum-free generation of hybridomas. The NS0 cell line is the property of the Medical Research Council (MRC). ECACC is unable to supply this cell line or derivatives of this cell line to commercial entities unless a license has been first secured from LifeArc (formerly known as MRC Technology). Requests for commercial use of this cell line will be forwarded to LifeArc.

Application

Fusion partner for generation of serum-free hybridomas.

Culture Medium

EX-CELL®NS0 Serum-Free Medium for NS0 Cells (Sigma 14650C) + 8mM L-Glutamine (G7513). When freezing cells down use EX-CELL®NS0 Serum-Free Medium for NS0 Cells (Sigma 14650C) + 8mM L-Glutamine (G7513) + 10% DMSO (D2650).

Subculture Routine

Viability may be poor on resuscitation and may initially decrease further. Full recovery may take up to 2 weeks. A centrifugation step to remove the cryoprotectant is essential. Rapidly thaw the frozen ampoule in a water bath at 37°C for 1-2 minutes. Transfer the contents to a centrifuge tube and slowly add 5-10ml of pre-warmed growth media. Remove a sample for counting. Centrifuge at 100g for 2-3 minutes to pellet cells and seed at a relatively high density of 5-7 x 105 cells/ml. Leave culture flask upright and observe regularly until viable proliferating cells are seen. Once established use a split ratio of 1:2 approximately every 4 to 5 days; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service