Skip to Content
MilliporeSigma

A9576

Bovine Serum Albumin solution

30% in DPBS, sterile-filtered, BioXtra, suitable for cell culture

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.75
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
28-32% total protein basis (UV)
Form:
liquid
Technique(s):
cell culture | mammalian: suitable
Impurities:
endotoxin, tested
Concentration:
in phosphate buffer (DPBS)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

sterility

sterile-filtered

product line

BioXtra

assay

28-32% total protein basis (UV)

form

liquid

origin

USA origin

concentration

in phosphate buffer (DPBS)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

pH

6.5-7.5

foreign activity

Mycoplasma Test by Barile on Raw material, none detected

shipped in

ambient

storage temp.

2-8°C

General description

BSA is a single polypeptide chain consisting of about 583 amino acid residues and no carbohydrates. At pH 5-7 it contains 17 intrachain disulfide bridges and 1 sulfhydryl group.

Application

BSA 30% solution is a convenient form for direct supplementation of serum-free media. Bovine serum albumin is broadly used as an additive to cell culture media, especially serum-free media. It provides a range of benefits including protection from oxidative damage and stabilization of other media components such as fatty acids and pyridoxal.

Preparation Note

Prepared from A9647 in DPBS (D5652).


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin solution


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library