Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SP2/O-Ag14 Cell Line murine

Non-producing hybridoma, 85072401

Synonym(s):

Mouse Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

mouse spleen

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse myeloma, non-producing

Cell Line Description

Sp2/0-Ag14 is a non-Ig-secreting or synthesising line derived from a cell line created by fusing a BALB/c mouse spleen cell and the mouse myeloma P3X63Ag8. Resistant to 8-azaguanine at 20ug/ml and does not survive in HAT containing media. HAT sensitive mouse myeloma hybrid suitable for use as a hybridoma fusion partner - in this sense, functionally it is a myeloma cell line and often referred to as such in the literature.

Application

Fusion partner

Culture Medium

RPMI 1640 or DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. HAT sensitive.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Eukaryotic cell lines aid in complex post-translational modifications and folding of recombinant proteins. Find suitable recombinant protein production cell lines.

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service