Skip to Content
MilliporeSigma
All Photos(1)

Documents

My-La CD4+

95051032, human lymph, Lymphoblastoid

Synonym(s):

My-La CD4+ Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

My-La CD4+, 95051032

biological source

human lymph

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian CD4+ T Lymphocyte, mycosis fungoides

Cell Line Description

Two different continuous cell lines were established from a plaque biopsy of an 82-year old male with mycosis fungoides stage II by inclusion of IL-2 and IL-4 in the culture medium: My-La, CD4+, (referred to in the literature as My-La, marker chromosome) and My-La, CD8+ (referred to in the literature as My-La 46,XY-18+i(18q) (Sigma Catalogue number. 95051033). My-La CD4+ is strictly dependent on exogenous IL-2 and IL-4 in the culture medium whereas My-La CD8+ on IL-2 only. Both show characteristic chromosome markers and are double homozygous for HLA-A1.B8. My-La CD8+ expresses β-13 of the T cell receptor and phenotypically weak staining was observed for CD2, CD3, CD8, CD25, CD71 and TCR-1 α / ß. My-La CD4+ expresses β-18 of the T cell receptor plus staining for CD2, CD3, CD4,CD25, CD71, HLA-DR and TCR-1 α/ ß. There has been no evidence of HTLV-I infection in the cell lines and serum from the patient did not demonstrate antibodies against HTLV-I and HTLV-II. Both cell lines show a low viability after resuscitation from frozen and a long lag time phase until exponential growth is reached.

Application

Study of antigen-independent proliferation of T- cell lyphoma from mycosis fungoides patient.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 11,12
D16S539: 8,11
D5S818: 9,12
D7S820: 10,11
THO1: 6
TPOX: 8,12
vWA: 17

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37°C. Split only 1:2 as maximum dilution: freeze in 45% growth medium, 45% human AB serum plus 10% DMSO, low viability on resuscitation. Use of human serum produces high levels of debris in cultures. Population doubling time 20-24 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service