Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1907

Sigma-Aldrich

Calcineurin from bovine brain

lyophilized powder, ≥2,500 units/mg protein

Synonym(s):

Calcium/Calmodulin-Activated Protein Phosphatase, Calmodulin binding protein, Modulator binding protein, PP2B, Phosphoprotein Phosphohydrolase, Protein phosphatase 2B

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

bovine

Quality Level

form

lyophilized powder

specific activity

≥2,500 units/mg protein

mol wt

dimer ~77 kDa
subunit mol wt 19-58 kDa

composition

Protein, 0.3-1.7% Lowry

solubility

H2O: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Calcineurin (CaN) comprises CaN A and CaN B subunits. It exists as a heterodimer with a calmodulin-binding domain, catalytic site, a CaN B binding domain, and an autoinhibitory domain.

Application

Calcineurin from bovine brain has been used:
  • as a positive control in western blot analysis of oocytes and cumulus cells proteome
  • as a positive control in calmodulin (CaM)-agarose binding assay
  • to test its phosphatase activity in the presence of okadaic acid

Biochem/physiol Actions

Calcineurin (CaN) activity is stimulated by nickel (Ni2+) and manganese (Mn2+) ions. It participates in the coupling of Ca2+ signals. CaN may regulate oocyte growth and meiotic maturation in porcine. It also participates in gene regulation, cell survival, and death.
Calcineurin is a cyclosporin-sensitive, calcium-regulated, serine-threonine protein phosphatase with broad substrate specificity. It is the major calmodulin-binding protein found in the brain. First identified as an inhibitor of the calmodulin activation of phosphodiesterase 3′:5′ cyclic nucleotide (PDE), calcineurin has similar effects on adenylate cyclase. Serves as a key enzyme involved in T-cell activation. Also involved in the hyperphosphorylation of tau protein in Alzheimer′s disease and has been shown to prevent calpain-mediated proteolysis of tau in differentiated PC12 cells.

Unit Definition

One unit will cause a 50% inhibition of the activated phosphodiesterase, 3′:5′-cyclic nucleotide (P 9529) activity when assayed with two units of activator (P 2277) and 0.1 mM Ca2+ in an enzyme coupled system at pH 7.5 at 30 °C.

Physical form

Lyophilized powder containing 0.5% EGTA, buffer salts and stabilizers.

Analysis Note

Sigma tests activity in 80 mM Tris, pH 7.5 with 65 mM KCl, 8 mM MgSO4 and 0.3% albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulator binding protein. Bovine brain protein exhibiting the Ca2+-dependent association with the protein modulator of cyclic nucleotide phosphodiesterase.
J H Wang et al.
The Journal of biological chemistry, 252(12), 4175-4184 (1977-06-25)
Ashakumary Lakshmikuttyamma et al.
Neurochemical research, 29(10), 1913-1921 (2004-11-10)
A major cause of neuronal dysfunction is due to altered Ca2+ regulation. An increase in Ca2+ influx can activate Ca2+-dependent enzymes including calpains, causing the proteolysis of its specific substrates. In the present study, calcineurin (CaN) was found to be
Calmodulin binds to and regulates the activity of beta-secretase (BACE1)
Chavez SE and O?Day DH
Current research journal of biological sciences, 1, 37-47 (2007)
C X Gong et al.
Brain research, 741(1-2), 95-102 (1996-11-25)
Abnormally hyperphosphorylated tau is the major protein component of neurofibrillary tangles, the characteristic lesion of Alzheimer's disease (AD). Protein phosphatases (PP) type 1 (PP-1), type 2A (PP-2A) and type 2B (PP-2B) appear to be involved in the regulation of tau
Felicia Ranta et al.
Cellular signalling, 20(10), 1780-1786 (2008-07-10)
Previously, we described that apoptotic cell death induced by the synthetic glucocorticoid dexamethasone (dex) is inhibited by calcineurin inhibitors, FK506 and deltamethrin, in insulin-secreting cells. The aim of the present study was to examine the mechanism of dex-dependent activation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service