Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0270

Sigma-Aldrich

Calmodulin from bovine heart

lyophilized powder, 2,500-10,000 units/mg protein

Synonym(s):

Phosphodiesterase 3′:5′-cyclic nucleotide activator from bovine heart

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

bovine heart

Quality Level

form

lyophilized powder

specific activity

2,500-10,000 units/mg protein

composition

Protein, ≥85% Lowry

technique(s)

calcium flux assay: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

Application

This is the protein activator required for the Ca2+ dependent activation of phosphodiesterase, 3′:5′-cyclic nucleotide, P0520.

Biochem/physiol Actions

Ca2+ binding protein that is required for activation of cyclic nucleotide-dependent phosphodiesterase. It is also a cofactor/activator of nitric oxide synthase, calcineurin, and many kinases including ATPase, myosin light chain kinase, and CAM kinase I, II, and III. It mediates ryanodine receptor activation by cyclic ADP ribose and is involved in intracellular Ca2+ homeostasis.

Unit Definition

One unit will stimulate 0.008 activated unit of phosphodiesterase, 3′:5′-cyclic nucleotide, P0520, in a 3 mL reaction volume at pH 7.5 and 30 °C, to 50% of the maximum activity of the enzyme when saturated with activator, in the presence of 0.01 mM Ca2+.

Physical form

Lyophilized powder containing buffer salts as imidazole and magnesium sulfate

Preparation Note

Purified by a modification of the procedure of Teo, T.S., et al., J. Biol. Chem., 248, 588 (1973).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service