Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

207008

Sigma-Aldrich

Calcineurin Substrate

An excellent peptide substrate for calcineurin.

Synonym(s):

Calcineurin Substrate, PP2B Substrate, RII Phosphopeptide, DLDVPIPGRFDRRVpSVAAE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C92H151N28O32P
Molecular Weight:
2192.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 2 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

An excellent peptide substrate for calcineurin (Protein Phosphatase 2B, PP2B, Cat. No. 539568). The sequence is derived from the PKA regulatory subunit type II (RII) and is phosphorylated on the Ser residue. Release of phosphate can be quantified colorimetrically using malachite green method.
An excellent peptide substrate for calcineurin (Protein Phosphatase 2B, PP2B; Cat. No. 539568). The sequence is derived from the PKA regulatory subunit type II (RII) and is phosphorylated on the Ser residue. Release of phosphate can be quantified colorimetrically using the malachite green method.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Calcineurin (Protein Phosphatase 2B, PP2B)
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

H-Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-Arg-Val-Ser(PO₃H₂)-Val-Ala-Ala-Glu-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Donella-Deana, A., et al. 1994. Eur. J. Biochem. 219, 109.
Enz, A., et al. 1994. Anal. Biochem. 216, 147.
Harder, K.W., et al. 1994. Biochem. J. 298, 395.
Fruman, D.A., et al. 1992. Proc. Natl. Acad. Sci. USA 89, 3686.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service