Skip to Content

P1868

Protein Phosphatase 2A2 from bovine kidney

buffered aqueous glycerol solution

Synonym(s):

PP2A2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
~2.0 U/vial
Biological source:
bovine kidney
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine kidney

Quality Segment

form

buffered aqueous glycerol solution

specific activity

~2.0 U/vial

mol wt

dimer 101 kDa (65 kDa and 36 kDa)

shipped in

dry ice

storage temp.

−70°C

General description

Protein phosphatase 2A (PP2A) is a specific protamine-kinase-inactivating phosphatase, one common physiological form of which is PP2A2.

Application

Protein phosphatase 2A2 has been used in a study to investigate two heat-stable protein inhibitors. It has also been used in a study to describe the purification and properties of a protamine kinase from bovine kidney microsomes.

Biochem/physiol Actions

Protein Phosphatase 2A2 from bovine kidney was shown to be a unique inhibitor of protamine kinase while other phosphatases in the same family including PP1, PP2B, PP2C did not show any inhibition.

Physical form

Solution of 1 μg per vial in 50 μl of 50 mM Tris-HCl, pH 7.0, containing 14 mM β-Mercaptoethanol, 1mM benzamidine, 0.1 mM PMSF, 1mM EDTA, and 50% glycerol.

Other Notes

One unit will release 1 nanomole of inorganic phosphate from (32)P-labeled phosphorylase per minute at pH 7.0 at 30 deg C.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library