Skip to Content
MilliporeSigma
All Photos(1)

Documents

KYSE-30

94072011, human esophagus, Polygonal

Synonym(s):

KYSE30 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

KYSE-30, 94072011

biological source

human esophagus

growth mode

Adherent

karyotype

Aneuploid

morphology

Polygonal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Asian squamous cell carcinoma

Cell Line Description

KYSE-30 was established from the oesophageal cancer of an untreated 64 year old male. The tumour sample was taken from the mucosal surface of a well differentiated squamous cell carcinoma. The cell line KYSE-30 was established with the use of tumours initially transplanted to athymic mice. The cells are reported to have a doubling time of 20.8 hrs in the exponential growth phase. A p53 mutation at the splice acceptor site of intron 6 and a 12 fold amplification of c-erb B has been reported. KYSE-30 cells express a large number of epidermal growth factor receptors, 1.2x10,000,000 sites/cell.

Application

Study of human oesophageal cancer.

Culture Medium

RPMI 1640 + Ham′s F12 (1:1) + 2mM Glutamine + 2% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 i.e. seeding at 1x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service