Skip to Content
MilliporeSigma
All Photos(1)

Documents

323381

Sigma-Aldrich

Hydrogen peroxide solution

contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:
NACRES:
NA.21

vapor density

1.1 (vs air)

Quality Level

vapor pressure

23.3 mmHg ( 30 °C)

form

liquid

contains

~200 ppm acetanilide as stabilizer

reaction suitability

reagent type: oxidant

concentration

3 wt. % in H2O
3.0-4.0 wt. % (H2O2 basis with KMnO4, redox titration)

refractive index

n20/D 1.335

density

1 g/mL at 25 °C

storage temp.

10-30°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogen peroxide solution is a significant commodity chemical with several uses in the fine chemical sector. H2O2 is also used in the chemical industry to produce fine and bulk compounds, as well as for the treatment of wastewater. It is an oxidizing agent, widely found in bleaching agents, cosmetics, detergents.

Application

Hydrogen peroxide solution is used as:
  • An oxidizing agent along with the urea in the epoxidation of propylene using TS-1(titanium silicate-1) catalyst.
  • An oxidizing agent in the oxidation of As(III) to As (V).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoichi Iriguchi et al.
Blood, 125(2), 370-382 (2014-10-29)
Although overexpression of T-bet, a master transcription factor in type-1 helper T lymphocytes, has been reported in several hematologic and immune diseases, its role in their pathogenesis is not fully understood. In the present study, we used transgenic model mice
Osamu Sakai et al.
Investigative ophthalmology & visual science, 56(1), 538-543 (2015-01-13)
The purpose of the present study was to investigate the role of glutathione peroxidase 4 (GPx4) in conjunctival epithelial cells. An immortalized human conjunctival epithelial cell line was used. Cells were transfected with catalase, GPx1, GPx4, SOD1, SOD2, or control
Koen van der Maaden et al.
Pharmaceutical research, 31(7), 1846-1854 (2014-01-29)
The aim of the study was to develop a cheap and fast method to produce hollow microneedles and an applicator for injecting vaccines into the skin at a pre-defined depth and test the applicability of the system for dermal polio
T Ahn et al.
Journal of neuroendocrinology, 27(2), 100-110 (2014-12-17)
Prenatal exposure of the female ovine foetus to excess testosterone leads to neuroendocrine disruptions in adulthood, as demonstrated by defects in responsiveness with respect to the ability of gonadal steroids to regulate gonadotrophin-releasing hormone (GnRH) secretion. In the ewe, neurones
Guilherme D Silva et al.
Journal of neuroinflammation, 12, 10-10 (2015-01-21)
Motor cortex stimulation (MCS) is an effective treatment in neuropathic pain refractory to pharmacological management. However, analgesia is not satisfactorily obtained in one third of patients. Given the importance of understanding the mechanisms to overcome therapeutic limitations, we addressed the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service