Skip to Content
MilliporeSigma
All Photos(2)

Documents

386790-M

Sigma-Aldrich

Hydrogen Peroxide

Synonym(s):

Hydrogen peroxide solution, H₂O₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
NACRES:
NA.21

vapor pressure

23.3 mmHg ( 30 °C)

Quality Level

Assay

29-32%

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

pH

2-4 (20 °C)

density

1.11 g/cm3 at 20 °C

cation traces

Fe: ≤0.5 ppm
NH4+: ≤5.0 ppm
heavy metals: ≤1.0 ppm

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Powerful oxidant. Induces apoptosis in cultured rat cortical neurons.

Application


  • Application of copper (I) selective ligands for PET imaging of reactive oxygen species through metabolic trapping.: This research explores the use of copper(I) ligands to image reactive oxygen species, including hydrogen peroxide, using PET imaging, which could have significant implications for diagnostic biochemistry (Tada et al., 2024).

Warning

Toxicity: Corrosive (L)

Physical form

30% solution, stabilized.

Other Notes

Whittemore, E.R., et al. 1994. NeuroReport5, 1485.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service