Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

216763

Sigma-Aldrich

Hydrogen peroxide solution

greener alternative

contains inhibitor, 30 wt. % in H2O, ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

vapor density

1 (vs air)

vapor pressure

23.3 mmHg ( 30 °C)

Assay

29.0-32.0% (ACS specification)

form

liquid

quality

stabilized

reaction suitability

reagent type: oxidant

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

30 wt. % in H2O

impurities

≤0.0006 meq/g Titr. acid

evapn. residue

≤0.002%

color

APHA: ≤10

pH

3-4 (30% in hydrogen peroxide)

pKa (25 °C)

11.6

density

1.11 g/cm3

anion traces

chloride (Cl-): ≤3 ppm
nitrate (NO3-): ≤2 ppm
phosphate (PO43-): ≤2 ppm
sulfate (SO42-): ≤5 ppm

cation traces

Fe: ≤0.5 ppm
NH4+: ≤5 ppm
heavy metals (as Pb): ≤1 ppm

greener alternative category

storage temp.

2-8°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogen peroxide solution is commonly used as a bleaching agent. It shows germicidal and sporicidal properties. H2O2 is a reactive oxygen species formed as a reaction product of various aerobic metabolic processes. Its ultra-trace quantification in the concentration range of 10-11mole/ml by a fluorometric method has been described.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. Click here for more information.

Application

Greener oxidant used for bromination of 1,3-dicarbonyl compounds, amide bond formation from amines and aldehydes, halogenation of aromatic compounds and dethioacetalization.

Highly Selective Oxidative Monochlorination To Synthesize Organic Intermediates: 2-Chlorotoluene, 2-Chloroaniline, 2-Chlorophenol, and 2-Chloro-4-methylphenol
Hydrogen peroxide solution has also been used in the preparation of ′Piranha′, a sulfuric acid/hydrogen peroxide mixture for cleaning silicon wafers used in nanostructure-initiator mass spectrometry (NIMS) chips.
It can be used as an oxidant for:
  • Epoxidation of terminal olefins under halide-free conditions.
  • Oxidation of cyclohexene to adipic acid under organic solvent- and halide-free conditions.
Hydrogen peroxide solution has been used in an immunoblotting protocol.

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A practical method for epoxidation of terminal olefins with 30% hydrogen peroxide under halide-free conditions.
Sato K, et al.
The Journal of Organic Chemistry, 61(23), 8310-8311 (1996)
Vapor-phase hydrogen peroxide as a surface decontaminant and sterilant.
Klapes NA and Vesley D. O. N. A. L. D.
Applied and Environmental Microbiology, 56(2), 503-506 (1990)
A "green" route to adipic acid: Direct oxidation of cyclohexenes with 30 percent hydrogen peroxide.
Sato K, et al.
Science, 281(5383), 1646-1647 (1998)
Changes in bovine enamel after treatment with a 30% hydrogen peroxide bleaching agent.
Hyun-Jin P, et al.
Dental Materials Journal, 23(4), 517-521 (2004)
Nanostructure-initiator mass spectrometry: a protocol for preparing and applying NIMS surfaces for high-sensitivity mass analysis.
Woo HK, et al.
Nature Protocols, 3(8), 1341-1349 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service