Skip to Content
MilliporeSigma
All Photos(1)

Documents

06858

Sigma-Aldrich

Celite® S

filter aid, dried, untreated

Synonym(s):

Diatomaceous earth, Diatomaceous silica, Kieselguhr

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
SB.52

form

powder

quality

dried
untreated

composition

Al2O3, 4.1%
CaO, 0.4%
Fe2O3, 1.6%
MgO, 0.2%
Na2O + K2O, 1.4%
P2O5, 0.3%
SiO2, 90.2%
TiO2, 0.2%

loss

3.2-10.0% loss on ignition, 900°C

pH

~7 (25 °C, 10% in aq. suspension)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Celite® S is composed of SiO2 and is also known as diatomite or diatomaceous earth. It is used as a filter aid for rapid filtration in an aqueous solution.

Application

Highly insoluble, inert material for use as a filtration aid.

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Goran G Andrić et al.
Journal of economic entomology, 105(2), 670-678 (2012-05-23)
Insecticidal potential of natural zeolites and diatomaceous earths originating from Serbia against Sitophilus oryzae (L.) and Tribolium castaneum (Herbst) was evaluated. Two natural zeolite formulations (NZ and NZ Modified) were applied to wheat at rates of 0.50, 0.75, and 1.0
Messaouda Safa et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 65(10), 1729-1737 (2012-05-02)
The adsorption of Cu(2+), Zn(2+), Cd(2+) and Pb(2+) ions from aqueous solution by Algerian raw diatomite was studied. The influences of different sorption parameters such as contact pH solution, contact time and initial metal ions concentration were studied to optimize
James F Meadow et al.
Microbial ecology, 65(1), 171-179 (2012-08-29)
Geothermal influenced soils exert unique physical and chemical limitations on resident microbial communities but have received little attention in microbial ecology research. These environments offer a model system in which to investigate microbial community heterogeneity and a range of soil
Heezoo Kim et al.
Anesthesia and analgesia, 115(2), 244-252 (2012-05-16)
The activated clotting time (ACT) is widely used for monitoring heparin anticoagulation during cardiac surgery. Celite-based ACT values are prolonged when aprotinin is administered. MDCO-2010, a novel serine protease inhibitor, is currently being evaluated as a possible alternative to aprotinin.
Tino Jaschinski et al.
Rapid communications in mass spectrometry : RCM, 27(1), 109-116 (2012-12-15)
Organic matrices are the state-of-the-art ionization mediators in Laser Desorption/Ionization Mass Spectrometry (LDI-MS). Despite improvements in understanding matrix chemistry, interfering matrix-related signals complicate the analysis. Surface-assisted LDI techniques like desorption/ionization on silicon (DIOS) or nanostructure initiator mass spectrometry (NIMS) provide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service