Skip to Content
MilliporeSigma
All Photos(2)

Documents

22140

Sigma-Aldrich

Celite® 545

filter aid, treated with sodium carbonate, flux calcined

Synonym(s):

Diatomaceous earth, flux-calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
SB.54

form

powder

quality

flux calcined
treated with sodium carbonate

composition

Al2O3, 1.0%
CaO, 6.73%
Fe2O3, 0.46%
Na2O + K2O, 1.88%
SiO2, 89.0%

median particle size

≤125.30 μm (71% (acc. to Cilas))

pH

>8.5 (25 °C, 10% in aq. suspension)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diatomaceous earth is made up of natural amorphous silica formed from the fossilized skeletons of diatoms. It is extracted, crushed, dried and calcined with flux material. DE is widely used as filler in construction materials, paints and as an anti-caking agent for agricultural chemicals. It has high water and oil absorption ability.

Application

Diatomaceous earth may be used as an adsorbent for column chromatography.

Caution

Not Intended for cGMP Parenteral Pharmaceutical Manufacturing

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immobilization of lipases on clay, Celite 545, diethylaminoethyl-, and carboxymethyl-Sephadex and their interesterification activity.
Ansari SA and Husain Q.
Biotechnol. Tech., 12(5), 381-384 (1998)
Lactose hydrolysis from milk/whey in batch and continuous processes by concanavalin A-Celite 545 immobilized Aspergillus oryzae ? galactosidase.
Ansari SA and Husain Q.
Food and Bioproducts Processing, 90(2), 351-359 (2012)
Direct immobilization of polyphenol oxidases on Celite 545 from ammonium sulphate fractionated proteins of potato (Solanum tuberosum).
Khan AA, et al.
Journal of Molecular Catalysis. B, Enzymatic, 40(1), 58-63 (2006)
Violeta Chitu et al.
Cell reports, 30(9), 3004-3019 (2020-03-05)
CSF-1R haploinsufficiency causes adult-onset leukoencephalopathy with axonal spheroids and pigmented glia (ALSP). Previous studies in the Csf1r+/- mouse model of ALSP hypothesized a central role of elevated cerebral Csf2 expression. Here, we show that monoallelic deletion of Csf2 rescues most behavioral deficits
Yueru Li et al.
The American journal of pathology, 190(9), 1971-1981 (2020-06-27)
Leakage of retinal blood vessels, which is an essential element of diabetic retinopathy, is driven by chronic elevation of vascular endothelial growth factor (VEGF). VEGF quickly relaxes the endothelial cell barrier by triggering signaling events that post-translationally modify pre-existing components

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service