Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

525243

Sigma-Aldrich

Celpure® P300

meets USP/NF testing specifications

Synonym(s):

Diatomaceous earth, calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
NA.21

Agency

meets USP/NF testing specifications

Quality Level

form

solid

specific activity

≤1 EU/g (endotoxin)

composition

SiO2, 98.65%

parameter

0.300 (Permeability(Darcy))

technique(s)

ion chromatography: suitable

impurities

≤0.2% water solubles
≤2.0% acid solubles
≤50 mg non-siliceous content

particle size

0.45-0.65 mm

cation traces

As: ≤10 ppm (leachable)
Pb: ≤10 ppm (leachable)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Celpure® P300 (Diatomaceous earth, DE) is natural amorphous silica formed from the fossilized skeletons of diatoms. It is extracted, crushed, dried and calcined with or without flux material. DE is widely used as filler in construction materials, paints and as an anti-caking agent for agricultural chemicals. It has high water and oil absorption ability. The potential toxicity of different samples of DE has been assessed. DE is also employed as an adsorbent for column chromatography.

Application

Celpure® P300 has been used used in the column chromatographic isolation and extraction of testosterone and corticosterone from freeze-dried water dragon plasma samples.
For high-purity, high-throughput processes

Legal Information

Product of Imerys Filtration Minerals
Celpure is a registered trademark of Advanced Minerals Corporation

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Troy A Baird et al.
Hormones and behavior, 66(2), 393-400 (2014-06-08)
Water dragons (Intellegama [Physignathus] lesueurii) are large (to >1m) agamid lizards from eastern Australia. Males are fiercely combative; holding a territory requires incessant displays and aggression against other males. If a dominant male is absent, injured or fatigued, another male
The global variability of diatomaceous earth toxicity: a physicochemical and in vitro investigation.
Nattrass C, et al.
Journal of Occupational Medicine and Toxicology (London, England), 10(1), 25-25 (2015)
Andrew Z Wang et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 189(2), 261-269 (2019-01-23)
Life-history trade-offs result from allocation of limited energetic resources to particular traits at the expense of others. When resources are scarce, some traits will take priority over others in the degree of their expression. For example, the current reproduction may
Induction of p53-independent growth inhibition in lung carcinoma cell A549 by gypenosides.
Journal of cellular and molecular medicine (2015).
Liu JS, et al.
Journal of Cellular and Molecular Medicine, 19(7), 1697-1709 (2015)
H Checkoway et al.
British journal of industrial medicine, 50(7), 586-597 (1993-07-01)
A cohort mortality study was conducted among workers from two plants in the diatomaceous earth mining and processing industry in California. Diatomaceous earth consists of the skeletal remains of diatoms. Exposure to amorphous (non-crystalline) and crystalline silica in the form

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service