Skip to Content
MilliporeSigma

1.02686

Celite® 281 Diatomaceous Earth

Celite® 281. CAS 68855-54-9, pH 10 (100 g/l, H2O, 20 °C).

Synonym(s):

Celite® 281

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
11101527
EC Index Number:
272-489-0
NACRES:
NB.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Celite® 281, Celite® 281. CAS 68855-54-9, pH 10 (100 g/l, H2O, 20 °C).

Quality Segment

technique(s)

HPLC: suitable

pH

10 (20 °C, 100 g/L in H2O)

mp

>450 °C

solubility

0.0037 g/L

density

2.36 g/cm3 at 20 °C

bulk density

300 kg/m3

storage temp.

no temp limit

SMILES string

[Li]CCCC

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

Analysis Note

Identity: passes test
Loss on ignition (980 °C): ≤ 2.0 %

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library