Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W455320

Sigma-Aldrich

cis-3-Nonenyl acetate

≥95%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
4553
UNSPSC Code:
12164502
Flavis number:
09.672
NACRES:
NA.21
grade:
FG
Halal
Kosher
food allergen:
no known allergens

grade

FG
Halal
Kosher

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

contains

synthetic α-tocopherol as stabilizer

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; herbaceous; pear; tropical

SMILES string

O(CC\C=C/CCCCC)C(=O)C

InChI

1S/C11H20O2/c1-3-4-5-6-7-8-9-10-13-11(2)12/h7-8H,3-6,9-10H2,1-2H3/b8-7-

InChI key

LRQNQAVSERJKNU-FPLPWBNLSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service