Skip to Content
MilliporeSigma
All Photos(1)

Documents

W466033

Sigma-Aldrich

DL-4-Hydroxy-3-methoxymandelic acid

≥95%, FG

Synonym(s):

α,4-Dihydroxy-3-methoxybenzeneacetic acid, VMA, Vanillomandelic acid, Vanillylmandelic acid, Vanilmandelic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH(OH)CO2H
CAS Number:
Molecular Weight:
198.17
FEMA Number:
4660
Beilstein:
2213227
MDL number:
UNSPSC Code:
12352106
Flavis number:
08.134
NACRES:
NA.21

grade

FG

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU 1334/2008& 178/2002
FDA 21 CFR 117

Assay

≥95%

mp

132-134 °C (dec.) (lit.)

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

sweet; vanilla

SMILES string

OC1=C(OC)C=C(C(O)C(O)=O)C=C1

InChI

1S/C9H10O5/c1-14-7-4-5(2-3-6(7)10)8(11)9(12)13/h2-4,8,10-11H,1H3,(H,12,13)

InChI key

CGQCWMIAEPEHNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service