Skip to Content
MilliporeSigma
All Photos(1)

Documents

W431433

Sigma-Aldrich

Phenethyl decanoate

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28O2
CAS Number:
Molecular Weight:
276.41
FEMA Number:
4314
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.685
NACRES:
NA.21

grade

FG

Quality Level

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

honey; floral; sweet

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service