Skip to Content
MilliporeSigma

SML3841

Nilotinib hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

4-Methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-benzamide hydrochloride hydrate, Nilotinib monohydrochloride monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H22F3N7O·HCl·H2O
CAS Number:
Molecular Weight:
583.99
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(C1=CC(NC(C2=CC=C(C(NC3=NC=CC(C4=CN=CC=C4)=N3)=C2)C)=O)=CC(N5C=NC(C)=C5)=C1)F.Cl.O

InChI

1S/C28H22F3N7O.ClH.H2O/c1-17-5-6-19(10-25(17)37-27-33-9-7-24(36-27)20-4-3-8-32-14-20)26(39)35-22-11-21(28(29,30)31)12-23(13-22)38-15-18(2)34-16-38;;/h3-16H,1-2H3,(H,35,39)(H,33,36,37);1H;1H2

InChI key

YCBPQSYLYYBPDW-UHFFFAOYSA-N

Biochem/physiol Actions

Nilotinib hydrochloride hydrate is an orally available, selective and potent ATP-competitive wild-type and mutant Bcr-Abl kinase inhibitor that is 10-30-fold more potent than imatinib. Nilotinib is used to treat Philadelphia chromosome (Ph+)-positive chronic myelogenous leukemia (CML). Nilotinib reduces midbrain Bcr-Abl autophosphorylation, amyloid-β levels, and neuronal loss, as well as improves autophagosome clearance and reverses cognitive deficits in the Tg2576 transgenic mouse model of Alzheimer’s disease.
Orally available, selective and potent ATP-competitive wild-type and mutant Bcr-Abl kinase inhibitor

Disclaimer

Hygroscopic


pictograms

Health hazardEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library