Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

S6547

Sigma-Aldrich

Sodium sulfate

BioXtra, ≥99.0%

Synonym(s):

Na2SO4, sodium sulphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
powder
solubility:
H2O: 1 M, clear, colorless

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥99.0%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.01% Insoluble matter

mp

884 °C (lit.)

solubility

H2O: 1 M, clear, colorless

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.001%

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teppei Adachi et al.
Inflammatory bowel diseases, 21(1), 31-39 (2014-12-02)
Expression of heat shock protein A4 (HSPA4, also called Apg-2), a member of the HSP110 family, is induced by several forms of stress. The physiological and pathological functions of HSPA4 in the intestine remain to be elucidated. We assessed HSPA4
Clément Cartier et al.
Water research, 47(2), 661-671 (2012-11-24)
Release of lead from 80% partially replaced service lines was compared to full lead service lines using harvested-stabilized lead pipes and field brass connectors. After more than a year of stabilization, lead release was consistent with field samples. Over the
Arseto Y Bagastyo et al.
Water research, 47(1), 242-250 (2012-11-10)
Reverse osmosis concentrate from wastewater reclamation contains biorefractory trace organic contaminants that may pose environmental or health hazard. Due to its high conductivity, electrochemical oxidation of brine requires low voltage which is energetically favourable. However, the presence of chloride ions
Roosmarijn E Vandenbroucke et al.
Nature reviews. Drug discovery, 13(12), 904-927 (2014-11-08)
Matrix metalloproteinases (MMPs) are zinc-dependent endopeptidases that form a family of 24 members in mammals. Evidence of the pathological roles of MMPs in various diseases, combined with their druggability, has made them attractive therapeutic targets. Initial drug discovery efforts focused
Jiao Zhao et al.
Angewandte Chemie (International ed. in English), 53(44), 11808-11812 (2014-09-12)
Porous films of p-type CuInS2, prepared by sulfurization of electrodeposited metals, are surface-modified with thin layers of CdS and TiO2. This specific porous electrode evolved H2 from photoelectrochemical water reduction under simulated sunlight. Modification with thin n-type CdS and TiO2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service