Skip to Content
MilliporeSigma

Skip To

1614807

USP

Sodium sulfate anhydrous

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Sodium sulfate, Disodium sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

sodium sulfate

manufacturer/tradename

USP

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium sulfate anhydrous USP reference standard intended for use in specified quality tests and assays.

Also used to prepare system suitability solutions for the assay of Sodium Monofluorophosphate by liquid chromatography coupled to conductivity with suppression detector.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S56407196313464
description

United States Pharmacopeia (USP) Reference Standard

description

≥99.0%, suitable for plant cell culture

description

tested according to Ph. Eur., anhydrous

description

puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)

grade

pharmaceutical primary standard

grade

-

grade

anhydrous

grade

puriss.

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

-

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

-

mp

884 °C (lit.)

mp

884 °C (lit.)

mp

884 °C (lit.)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Sodium sulfate anhydrous


Storage Class

13 - Non Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hello, I was wondering if this sodium sulfate was of USP grade? Thanks!

    1 answer
    1. Product 1614807 is an analytical standard provided by the USP. It is tested according to the USP, but it is not intended for human or therapeutic use.

      Helpful?

Reviews

No rating value

Active Filters