Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

238597

Sigma-Aldrich

Sodium sulfate

Sodium sulfate
1 of 1 reviewers received a sample product or took part in a promotion

ACS reagent, ≥99.0%, anhydrous, powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Quality Level

Assay

≥99.0%

form

powder

impurities

≤0.01% insolubles
≤5 ppm N compounds

loss

≤0.5% loss on ignition

pH

5.2-9.2 (25 °C, 5% in water)
5.2-9.2

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

Ca: ≤0.002%
Fe: ≤0.001%
K: ≤0.002%
Mg: ≤0.001%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium sulfate can be used as a catalyst:
  • In the Suzuki-Miyaura cross-coupling reactions of aryl halides with arylboronic acids to synthesize biaryl compounds in the presence of PdCl2.
  • For the aminolytic depolymerization of PET (polyethylene terephthalate) bottle waste.
It can also be used as an additive in the Pd-catalyzed Sonogashira reaction of terminal alkynes with aryl halides to synthesize polyfunctional alkynes.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 405-405 (1994)
The use of sodium sulfate as the globulin precipitant in the determination of proteins in blood.
Howe PE.
The Journal of Biological Chemistry, 49(1), 93-107 (1921)
How does sodium sulfate crystallize? Implications for the decay and testing of building materials.
Rodriguez-Navarro C, et al.
Cement and Concrete Research, 30(10), 1527-1534 (2000)
Teppei Adachi et al.
Inflammatory bowel diseases, 21(1), 31-39 (2014-12-02)
Expression of heat shock protein A4 (HSPA4, also called Apg-2), a member of the HSP110 family, is induced by several forms of stress. The physiological and pathological functions of HSPA4 in the intestine remain to be elucidated. We assessed HSPA4
Clément Cartier et al.
Water research, 47(2), 661-671 (2012-11-24)
Release of lead from 80% partially replaced service lines was compared to full lead service lines using harvested-stabilized lead pipes and field brass connectors. After more than a year of stabilization, lead release was consistent with field samples. Over the

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    0 answers

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service