Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7309

Supelco

Sigma Pseudo Narcotic Scent Marijuana formulation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

format

neat

Quality Level

storage temp.

−20°C

Application

Sigma pseudo narcotic scent marijuana formulation may be used as a surrogate in the comparative study of odor profiles of real marijuana odor vs surrogate marijuana scent using multidimensional gas chromatography coupled with mass spectrometry-olfactometry (MDGC– MS-O).
Substitute for controlled substances in the training of narcotics detector dogs. Mimics the odor of marijuana.

Legal Information

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The relationship between chemical concentration and odor activity value explains the inconsistency in making a comprehensive surrogate scent training tool representative of illicit drugs
Rice S and Koziel A J.
Forensic Science International, 257, 257-270 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service