Skip to Content
MilliporeSigma
All Photos(1)

Documents

RAB0038

Sigma-Aldrich

Cocaine- and Amphetamine-responsive Transcript EIA Kit

for serum, plasma, culture supernatant and cell lysates

Synonym(s):

CART

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.32

species reactivity

human, rat, mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type plasma
sample type serum
sample type cell lysate
sample type culture supernatant(s)

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 13.5 pg/mL
standard curve range: 0.1-1000 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CARTPT(9607)

General description

The CART Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting CART peptide based on the principle of Competitive Enzyme Immunoassay.

Immunogen

Cocaine- and Amphetamine-responsive Transcript (CART), synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service