Skip to Content
MilliporeSigma
All Photos(1)

Documents

PSCI

Supelco

Sigma Pseudo Corpse Scent

Corpse Scent Kit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

format

neat

Looking for similar products? Visit Product Comparison Guide

Application

Canine training aids for the detection of corpses.

Packaging

Two applications each of Formulation I and Formulation II

Legal Information

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service