Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1547

Peach leaves

NIST® SRM® 1547

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in validating analytical methods for the determination of major, minor, and trace elements in botanical materials, agricultural food products, and similar materials. This SRM can also be used for quality assurance when assigning values to in-house control materials. A unit of SRM 1547 consists of 50 grams of dried peach leaves of the Coronet variety. For more information, please refer to the COA and SDS.

SRM 1547_cert

SRM 1547_SDS

Application

Peach leaves has been used as standard reference material
  • for determination of trace element in coffee by inductively coupled plasma mass spectrometry (ICP-MS)
  • for determination of selenium by hydride generation atomic fluorescence spectrometry (HG-AFS) in vegetables
  • to test the efficacy of microwave/ultrasound (MW/US)-assisted digestion method for the estimation of Kjeldahl nitrogen

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Nitrogen (N), Phosphorous
Trace Elements: Aluminum (Al), Antimony (SB), Barium (Ba), Boron (B), Bromine (Br), Cadmium (Cd), Calcium (Ca), Cerium (Ce), Chlorine (Cl2), Chromium (Cr), Cobalt (Co), Copper (Cu), Europium (Eu), Gadolinium (Gd), Iodine (I), Iron (Fe), Lanthanum (La), Lead (Pb), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Molybdenum (Mo), Neodymium (Nd), Nickel (Ni), Potassium (K), Rubidium (Rb), Samarium (Sm), Scandium (Sc), Sodium (Na), Strontium (Sr), Sulfur (S), Terbium (Tb), Thorium (Th), Uranium (U), Vanadium (V), Zinc Zn)

Matrix Group: Fruits and Vegetables

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of selenium in vegetables by hydride generation atomic fluorescence spectrometry
Smrkolj P and Stibilj V
Analytica Chimica Acta, 512(1), 11-17 (2004)
The use of advanced chemometric techniques and trace element levels for controlling the authenticity of organic coffee
Barbosa RM, et al.
Food Research International, 61, 246-251 (2014)
A simultaneous, direct microwave/ultrasound-assisted digestion procedure for the determination of total Kjeldahl nitrogen.
Domini C, et al.
Ultrasonics Sonochemistry, 16(4), 564-569 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service